(±)-Tetrahydrofurfuryl bromide ≥95.0%

Leverantör: TCI
Varning

Synonyms: 2-(Bromomethyl)tetrahydrofuran

T1808-25G T1808-500G T1808-100G
TCIAT1808-25GEA 405 SEK
TCIAT1808-25G TCIAT1808-500G TCIAT1808-100G
(±)-Tetrahydrofurfuryl bromide ≥95.0%
(±)-Tetrahydrofurfuryl bromide
Formel: C₅H₉BrO
Kokpunkt: 70°C
Flampunkt: 73°C
Storage Temperature: Kylskåp
MDL Number: MFCD00016894
CAS nummer: 1192-30-9
EINECS: 214-750-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82070b63e117a983f9c2be0314bb20c3ba3b3ffd&vid=82070b63e117a983f9c2be0314bb20c3ba3b3ffd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR