(+)-Di-p-toluoyl-D-tartaric acid ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: Di-p-toluoyl-D-tartaric acid, (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric Acid, Di-p-toluoyl-D-tartaric Acid

D1417-25G D1417-250G
TCIAD1417-25GEA 495 SEK
TCIAD1417-25G TCIAD1417-250G
(+)-Di-p-toluoyl-D-tartaric acid ≥98.0% (by HPLC, titration analysis)
(+)-Di-p-toluoyl-D-tartaric acid
Formel: C₂₀H₁₈O₈
MW: 386,36 g/mol
MDL Number: MFCD00008552
CAS nummer: 32634-68-7
EINECS: 251-132-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c46f0b55858bea3fdc79b3c28d9f6aadae05cde5&vid=c46f0b55858bea3fdc79b3c28d9f6aadae05cde5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR