N-Acetyl-β-D-glucosamine ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Synonyms:
2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine
A0092-25G
A0092-500G
A0092-100G
TCIAA0092-25GEA
245
SEK
TCIAA0092-25G
TCIAA0092-500G
TCIAA0092-100G
N-Acetyl-β-D-glucosamine ≥98.0% (by HPLC, total nitrogen)
N-Acetyl-β-D-glucosamine
Formel:
C₈H₁₅NO₆ MW: 221,21 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00136044 CAS nummer: 7512-17-6 EINECS: 231-368-2 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) (CAD) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | +39.0 to +42.0 deg (C=1, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...