Methyl tiglate ≥98.0%
Leverantör: TCI
|
Varning
|
T0248-25ML
T0248-100ML
TCIAT0248-25MLEA
685
SEK
TCIAT0248-25ML
TCIAT0248-100ML
Methyl tiglate ≥98.0%
Methyl tiglate
Formel:
C₆H₁₀O₂ Kokpunkt: 139°C Flampunkt: 39°C |
MDL Number:
MFCD00016654 CAS nummer: 6622-76-0 EINECS: 229-575-8 UN: 3272 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...