1,3-Dimethylimidazolium chloride ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,3-Dimethyl-1H-imidazol-3-ium chloride

D3341-25G D3341-5G
TCIAD3341-25GEA 8875 SEK
TCIAD3341-25G TCIAD3341-5G
1,3-Dimethylimidazolium chloride ≥98.0%
1,3-Dimethylimidazolium chloride
Formel: C₅H₉ClN₂
MW: 132,5900 g/mol
Smältpunkt: 125°C
MDL Number: MFCD08276366
CAS nummer: 79917-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=364c5b53227b588da767aa1bde8ec229dd9a94f6&vid=364c5b53227b588da767aa1bde8ec229dd9a94f6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR