3-Methylglutaric acid ≥99.0%
Leverantör: TCI EUROPE
|
Varning
|
M0330-25G
M0330-5G
TCIAM0330-25GEA
2475
SEK
TCIAM0330-25G
TCIAM0330-5G
3-Methylglutaric acid ≥99.0%
3-Methylglutaric acid
Formel:
C₆H₁₀O₄ MW: 146,1412 g/mol |
MDL Number:
MFCD00002727 CAS nummer: 626-51-7 EINECS: 210-951-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 85.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...