Cyclohexyl isocyanide ≥98.0%

Leverantör: TCI
Fara

C1438-5G C1438-1G
TCIAC1438-5GEA 1525 SEK
TCIAC1438-5G TCIAC1438-1G
Cyclohexyl isocyanide ≥98.0%
Cyclohexyl isocyanide
Formel: C₇H₁₁N
Kokpunkt: 78°C
Flampunkt: 77°C
MDL Number: MFCD00003839
CAS nummer: 931-53-3
EINECS: 213-238-7
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1361acc9db07e0721e2f9fe081985fe381bcb654&vid=1361acc9db07e0721e2f9fe081985fe381bcb654&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR