1-Iodoisoquinoline ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
I0750-25G
I0750-5G
TCIAI0750-25GEA
7825
SEK
TCIAI0750-25G
TCIAI0750-5G
1-Iodoisoquinoline ≥98.0% (by GC, titration analysis)
1-Iodoisoquinoline
Formel:
C₉H₆IN |
MDL Number:
MFCD00234494 CAS nummer: 19658-77-6 |
|
Specifikation testresultat
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 73.0 to 77.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...