4-(Benzyloxy)-3-methoxybenzaldehyde ≥98.0%
Leverantör: TCI
B3599-25G B3599-5G
TCIAB3599-25GEA 1825 SEK
TCIAB3599-25G TCIAB3599-5G
4-(Benzyloxy)-3-methoxybenzaldehyde ≥98.0%
4-(Benzyloxy)-3-methoxybenzaldehyde
Formel: C₁₅H₁₄O₃
Kokpunkt: 185°C
MDL Number: MFCD00003365
CAS nummer: 2426-87-1
EINECS: 219-379-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f0d8d77d06bc7b64c5fcd913e3443871b767f09&vid=2f0d8d77d06bc7b64c5fcd913e3443871b767f09&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C