(S)-(+)-Prolinol ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: (S)-(+)-2-Hydroxymethylpyrrolidine, (2S)-pyrrolidin-2-ylmethanol, L(+)-Prolinol, S(+)-2-Pyrrolidinemethanol, L-Prolinol
P1087-25G P1087-5G
TCIAP1087-25GEA 2150 SEK
TCIAP1087-25G TCIAP1087-5G
(S)-(+)-Prolinol ≥97.0% (by GC, titration analysis)
(S)-(+)-Prolinol
Formel: C₅H₁₁NO
MW: 101,15 g/mol
Kokpunkt: 66°C
Flampunkt: 86°C
MDL Number: MFCD00005255
CAS nummer: 23356-96-9
EINECS: 245-605-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b66aa29f6402b50cae69c3561298f8dbe15a9ca&vid=0b66aa29f6402b50cae69c3561298f8dbe15a9ca&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D +28.0 to +33.0 deg (C=1,Toluene)