3-Methyl-3-oxetanemethanol ≥96.0%
Leverantör: TCI EUROPE
Synonyms: 3-Methyl-3-oxethanemethanol
M1136-25G M1136-5G
TCIAM1136-25GEA 685 SEK
TCIAM1136-25G TCIAM1136-5G
3-Methyl-3-oxetanemethanol ≥96.0%
3-Methyl-3-oxetanemethanol
Formel: C₅H₁₀O₂
MW: 102,13 g/mol
Kokpunkt: 122°C
Flampunkt: 97°C
Storage Temperature: Kylskåp
MDL Number: MFCD00010273
CAS nummer: 3143-02-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b22cd8f5ece66e206bbca4d53860f9c5aa72da&vid=f6b22cd8f5ece66e206bbca4d53860f9c5aa72da&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %