5-Bromoindoline ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
B3885-1G B3885-5G
TCIAB3885-1GEA 365 SEK
TCIAB3885-1G TCIAB3885-5G
5-Bromoindoline ≥98.0% (by GC, titration analysis)
5-Bromoindoline
Formel: C₈H₈BrN
Kokpunkt: 132°C
Storage Temperature: Frys
MDL Number: MFCD00027410
CAS nummer: 22190-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a52b883305d3d98bea6633dbe02210988114bfd&vid=1a52b883305d3d98bea6633dbe02210988114bfd&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 39.0 to 43.0 deg-C