Triethyl phosphonoacetate ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Ethyl 2-(diethoxyphosphoryl)acetate, Diethyl [(ethoxycarbonyl)methyl]phosphonate

D1523-25G D1523-500G D1523-100G
TCIAD1523-25GEA 215 SEK
TCIAD1523-25G TCIAD1523-500G TCIAD1523-100G
Triethyl phosphonoacetate ≥97.0%
Triethyl phosphonoacetate
Formel: C₈H₁₇O₅P
MW: 224,19 g/mol
Kokpunkt: 145°C
Flampunkt: 165°C
MDL Number: MFCD00009177
CAS nummer: 867-13-0
EINECS: 212-757-6
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e83a5e84083bf415312380abfbab958acf973714&vid=e83a5e84083bf415312380abfbab958acf973714&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR