1-Bromoisoquinoline ≥98.0%

Leverantör: TCI
Varning

B3445-1G B3445-5G
TCIAB3445-1GEA 2400 SEK
TCIAB3445-1G TCIAB3445-5G
1-Bromoisoquinoline ≥98.0%
1-Bromoisoquinoline
Formel: C₉H₆BrN
MDL Number: MFCD00234478
CAS nummer: 1532-71-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29f92407f080517465e0c4cd7f68bdb36c154d90&vid=29f92407f080517465e0c4cd7f68bdb36c154d90&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR