4-Nitro-p-terphenyl 95,0 GC_ANALYSMETOD
Leverantör: TCI
N0610-1G
N0610-5G
TCIAN0610-1GEA
710
SEK
TCIAN0610-1G
TCIAN0610-5G
4-Nitro-p-terphenyl 95,0 GC_ANALYSMETOD
4-Nitro-p-terphenyl
Formel:
C₁₈H₁₃NO₂ |
MDL Number:
MFCD00044846 CAS nummer: 10355-53-0 EINECS: 233-775-0 |
|
Specifikation testresultat
Purity (GC) | min. 95.0 % |
Melting point | 211.0 to 216.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...