(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol 98,0 GC_ANALYSMETOD
Leverantör: TCI
|
Varning
|
Synonyms:
(S)-1,2,3,4-Tetrahydronaphthalen-1-ol, (S)-(+)-alpha-Tetralol
T2360-1G
T2360-100MG
TCIAT2360-1GEA
7150
SEK
TCIAT2360-1G
TCIAT2360-100MG
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol 98,0 GC_ANALYSMETOD
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol
Formel:
C₁₀H₁₂O MW: 148.2 g/mol Kokpunkt: 92°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00063006 CAS nummer: 53732-47-1 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 39.0 to 43.0 deg-C |
Specific rotation [a]20/D | +30.0 to +36.0 deg (C=2.5, CHCl3) |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...