Chloromethyl methyl sulphide ≥97.0%
Leverantör: TCI EUROPE
Fara
C0608-25G C0608-100G
TCIAC0608-25GEA 1125 SEK
TCIAC0608-25G TCIAC0608-100G
Chloromethyl methyl sulphide ≥97.0%
Chloromethyl methyl sulphide
Formel: C₂H₅ClS
MW: 96,58 g/mol
Kokpunkt: 106°C
Flampunkt: 17°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000923
CAS nummer: 2373-51-5
EINECS: 219-148-4
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e17eda787bea9384d346b6ada8f019e05d2985c8&vid=e17eda787bea9384d346b6ada8f019e05d2985c8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %