Dimethylphenylsilane ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Dimethyl(phenyl)silane
D2196-25ML
D2196-5ML
TCIAD2196-25MLEA
1575
SEK
TCIAD2196-25ML
TCIAD2196-5ML
Dimethylphenylsilane ≥97.0%
Dimethylphenylsilane
Formel:
C₈H₁₂Si MW: 136,27 g/mol Kokpunkt: 158°C Flampunkt: 35°C |
MDL Number:
MFCD00008257 CAS nummer: 766-77-8 EINECS: 212-170-5 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...