2-Iodohippuric acid ≥99.0%
Leverantör: TCI EUROPE
Synonyms:
N-(2-Iodobenzoyl)glycine
I0446-25G
I0446-5G
TCIAI0446-25GEA
2475
SEK
TCIAI0446-25G
TCIAI0446-5G
2-Iodohippuric acid ≥99.0%
2-Iodohippuric acid
Formel:
C₉H₈INO₃ MW: 305,07 g/mol |
MDL Number:
MFCD00002694 CAS nummer: 147-58-0 EINECS: 205-693-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 173.0 to 177.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...