Triethylenetetramine >60.0%

Leverantör: TCI EUROPE
Fara

Synonyms: N,N'-Bis(2-aminoethyl)-1,2-ethanediamine, Trientine, DEH 24, EPH 925

T0429-25ML T0429-500ML
TCIAT0429-25MLEA 230 SEK
TCIAT0429-25ML TCIAT0429-500ML
Triethylenetetramine >60.0%
Triethylenetetramine
Formel: C₆H₁₈N₄
MW: 146,24 g/mol
Kokpunkt: 266°C
Flampunkt: 143°C
MDL Number: MFCD00008169
CAS nummer: 112-24-3
EINECS: 203-950-6
UN: 2259
ADR: 8,II
Merck Index: 14,09663
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbaca22a2357d563d08c97b56770c8899b5bcb87&vid=cbaca22a2357d563d08c97b56770c8899b5bcb87&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 60.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR