(4-Pyridylthio)acetic acid ≥98.0%

Leverantör: TCI
Varning

Synonyms: 2-(Pyridin-4-ylthio)acetic acid

P1000-25G P1000-500G
TCIAP1000-25GEA 325 SEK
TCIAP1000-25G TCIAP1000-500G
(4-Pyridylthio)acetic acid ≥98.0%
(4-Pyridylthio)acetic acid
Formel: C₇H₇NO₂S
MDL Number: MFCD00006424
CAS nummer: 10351-19-6
EINECS: 233-765-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1e1891d5dbbe496de1922e609dc8ee725c5cef2&vid=f1e1891d5dbbe496de1922e609dc8ee725c5cef2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in 0.5mol/L NaOH very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR