(2,4-Dichlorophenyl)acetone ≥98.0%

Leverantör: TCI

Synonyms: 1-(2,4-Dichlorophenyl)-2-propanone, 2,4-Dichlorophenylacetone

D2928-25G D2928-5G
TCIAD2928-25GEA 6225 SEK
TCIAD2928-25G TCIAD2928-5G
(2,4-Dichlorophenyl)acetone ≥98.0%
(2,4-Dichlorophenyl)acetone
Formel: C₉H₈Cl₂O
MDL Number: MFCD00210405
CAS nummer: 93457-07-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b18650974e208c262c182f5306bc1cac66c60e02&vid=b18650974e208c262c182f5306bc1cac66c60e02&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Melting point 46.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR