Diethyl methylmalonate ≥98.0%
Leverantör: TCI
Synonyms:
Diethyl 2-methylmalonate
M0366-25ML
M0366-500ML
M0366-100ML
TCIAM0366-25MLEA
215
SEK
TCIAM0366-25ML
TCIAM0366-500ML
TCIAM0366-100ML
Diethyl methylmalonate ≥98.0%
Diethyl methylmalonate
Formel:
C₈H₁₄O₄ Kokpunkt: 199°C Flampunkt: 82°C |
MDL Number:
MFCD00009162 CAS nummer: 609-08-5 EINECS: 210-175-7 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...