2-Ethyl-2-methyl-1,3-dioxolane ≥98.0%

Leverantör: TCI EUROPE
Fara

E0515-25ML E0515-500ML
TCIAE0515-25MLEA 4200 SEK
TCIAE0515-25ML TCIAE0515-500ML
2-Ethyl-2-methyl-1,3-dioxolane ≥98.0%
2-Ethyl-2-methyl-1,3-dioxolane
Formel: C₆H₁₂O₂
MW: 116,16 g/mol
Kokpunkt: 117°C
Flampunkt: 13°C
MDL Number: MFCD00014108
CAS nummer: 126-39-6
EINECS: 204-785-2
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d874e0b6a2285b23d03e45500e045c2f2eb4737c&vid=d874e0b6a2285b23d03e45500e045c2f2eb4737c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 0.9340 to 0.9440

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR