2-Methylbenzo[b]thiophene ≥98.0%

Leverantör: TCI

M1429-1G M1429-5G
TCIAM1429-1GEA 605 SEK
TCIAM1429-1G TCIAM1429-5G
2-Methylbenzo[b]thiophene ≥98.0%
2-Methylbenzo[b]thiophene
Formel: C₉H₈S
MDL Number: MFCD00216250
CAS nummer: 1195-14-8
EINECS: 214-792-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b869bf7c8a5d5ccba7909aa4d1f36ed6c173f4&vid=c2b869bf7c8a5d5ccba7909aa4d1f36ed6c173f4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR