N-Allylrhodanine ≥98.0%

Leverantör: TCI
Varning

Synonyms: 3-Allylrhodanine, 3-Allyl-2-thioxo-4-thiazolidinone

A1412-25G A1412-5G
TCIAA1412-25GEA 2850 SEK
TCIAA1412-25G TCIAA1412-5G
N-Allylrhodanine ≥98.0%
N-Allylrhodanine
Formel: C₆H₇NOS₂
MW: 173.26 g/mol
Kokpunkt: 139°C
MDL Number: MFCD00044700
CAS nummer: 1457-47-2
EINECS: 215-941-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=828700c7b90fb113474a5f480afff680bba47c8c&vid=828700c7b90fb113474a5f480afff680bba47c8c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR