2-Ethyl-2-oxazoline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
E0373-25G
E0373-500G
E0373-100G
TCIAE0373-25GEA
215
SEK
TCIAE0373-25G
TCIAE0373-500G
TCIAE0373-100G
2-Ethyl-2-oxazoline ≥98.0% (by GC, titration analysis)
2-Ethyl-2-oxazoline
Formel:
C₅H₉NO MW: 99,13 g/mol Kokpunkt: 128°C Flampunkt: 32°C |
MDL Number:
MFCD00005302 CAS nummer: 10431-98-8 EINECS: 233-912-4 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...