Phthalylsulphathiazole ≥95.0%

Leverantör: TCI

Synonyms: Phthalylsulphathiazole, 2-((4-(N-(Thiazol-2-yl)sulphamoyl)phenyl)carbamoyl)benzoic acid, 4'-(2-Thiazolylsulphamoyl)phthalanilic acid

T0703-25G T0703-500G
TCIAT0703-25GEA 215 SEK
TCIAT0703-25G TCIAT0703-500G
Phthalylsulphathiazole ≥95.0%
Phthalylsulphathiazole
Formel: C₁₇H₁₃N₃O₅S₂
MW: 403.44
MDL Number: MFCD00005318
CAS nummer: 85-73-4
EINECS: 201-627-4
Merck Index: 14,07377
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efdde6b8cb7050331fa6989d9823e450cc97b587&vid=efdde6b8cb7050331fa6989d9823e450cc97b587&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR