Bis(phenylthio)methane ≥97.0%

Leverantör: TCI EUROPE
Varning

B1444-25G B1444-5G
TCIAB1444-25GEA 1025 SEK
TCIAB1444-25G TCIAB1444-5G
Bis(phenylthio)methane ≥97.0%
Bis(phenylthio)methane
Formel: C₁₃H₁₂S₂
MW: 232,37 g/mol
Kokpunkt: 194°C
MDL Number: MFCD00003067
CAS nummer: 3561-67-9
EINECS: 222-624-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef1ede72f26ca1c284a9791150d6a8856ef9e757&vid=ef1ede72f26ca1c284a9791150d6a8856ef9e757&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 36.0 to 38.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR