Tributyl acetylcitrate ≥97.0%
Leverantör: TCI EUROPE
Synonyms:
Tributyl O-Acetylcitrate, Acetyltributylcitrate, Acetyl tributyl citrate, Tributyl 2-acetoxypropane-1,2,3-tricarboxylate, O-Acetylcitric acid tributyl ester
A0822-25ML
A0822-500ML
TCIAA0822-25MLEA
215
SEK
TCIAA0822-25ML
TCIAA0822-500ML
Tributyl acetylcitrate ≥97.0%
Tributyl acetylcitrate
Formel:
C₂₀H₃₄O₈ MW: 402,49 g/mol Smältpunkt: –80°C Flampunkt: 204°C |
MDL Number:
MFCD00043554 CAS nummer: 77-90-7 EINECS: 201-067-0 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...