O-Phenyl chlorothioformate ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Phenyl chlorothionocarbonate, Phenyl chlorothionoformate, O-Phenyl carbonochloridothioate
P1236-25G
P1236-5G
TCIAP1236-25GEA
1900
SEK
TCIAP1236-25G
TCIAP1236-5G
O-Phenyl chlorothioformate ≥98.0% (by GC, titration analysis)
O-Phenyl chlorothioformate
Formel:
C₇H₅ClOS MW: 172,63 g/mol Kokpunkt: 93°C Flampunkt: 81°C |
MDL Number:
MFCD00004920 CAS nummer: 1005-56-7 EINECS: 213-736-4 UN: 3265 ADR: 8,II |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...