1,3,5-Trimethylhexahydro-1,3,5-triazine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
T1422-25ML T1422-500ML
TCIAT1422-25MLEA 390 SEK
TCIAT1422-25ML TCIAT1422-500ML
1,3,5-Trimethylhexahydro-1,3,5-triazine ≥98.0% (by GC, titration analysis)
1,3,5-Trimethylhexahydro-1,3,5-triazine
Formel: C₆H₁₅N₃
Kokpunkt: 65°C
Flampunkt: 49°C
MDL Number: MFCD00006057
CAS nummer: 108-74-7
EINECS: 203-612-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ed7b73a52eee9672d43ef491346eef0410084af&vid=7ed7b73a52eee9672d43ef491346eef0410084af&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %