Nortropine ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
(3-endo)-8-Azabicyclo[3.2.1]octan-3-ol, 8-Azabicyclo[3.2.1]octan-3-ol
N0843-25G
N0843-5G
TCIAN0843-25GEA
5075
SEK
TCIAN0843-25G
TCIAN0843-5G
Nortropine ≥98.0%
Nortropine
Formel:
C₇H₁₃NO MW: 127.19 g/mol |
MDL Number:
MFCD00047140 CAS nummer: 538-09-0 |
|
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...