5-Bromo-2-methoxypyrimidine ≥98.0%
Leverantör: TCI EUROPE
Varning
B3521-1G B3521-5G
TCIAB3521-1GEA 605 SEK
TCIAB3521-1G TCIAB3521-5G
5-Bromo-2-methoxypyrimidine ≥98.0%
5-Bromo-2-methoxypyrimidine
Formel: C₅H₅BrN₂O
MW: 189,01 g/mol
MDL Number: MFCD04973351
CAS nummer: 14001-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=167f5f4dea35c7b3d407455b4928c246d3968721&vid=167f5f4dea35c7b3d407455b4928c246d3968721&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C