2,5-Piperazinedione ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Glycine anhydride, Piperazine-2,5-dione

G0100-25G G0100-500G
TCIAG0100-25GEA 550 SEK
TCIAG0100-25G TCIAG0100-500G
2,5-Piperazinedione ≥98.0%
2,5-Piperazinedione
Formel: C₄H₆N₂O₂
MW: 114,1 g/mol
MDL Number: MFCD00006009
CAS nummer: 106-57-0
EINECS: 203-411-5
Merck Index: 14,07466
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4536299a0bd9595261ae6cb71d00a8de0185c0a0&vid=4536299a0bd9595261ae6cb71d00a8de0185c0a0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR