Diethyl phenyl orthoformate ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
O0187-25G
O0187-5G
TCIAO0187-25GEA
2925
SEK
TCIAO0187-25G
TCIAO0187-5G
Diethyl phenyl orthoformate ≥97.0%
Diethyl phenyl orthoformate
Formel:
C₁₁H₁₆O₃ MW: 196,25 g/mol Kokpunkt: 104°C Flampunkt: 65°C |
MDL Number:
MFCD00009228 CAS nummer: 14444-77-0 EINECS: 238-421-9 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...