7-Tetradecyne 97,0 GC_ANALYSMETOD
Leverantör: TCI
|
Fara
|
T0762-25ML
T0762-5ML
TCIAT0762-25MLEA
6425
SEK
TCIAT0762-25ML
TCIAT0762-5ML
7-Tetradecyne 97,0 GC_ANALYSMETOD
7-Tetradecyne
Formel:
C₁₄H₂₆ MW: 194.36 g/mol Kokpunkt: 144°C |
MDL Number:
MFCD00041662 CAS nummer: 35216-11-6 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...