Dihydrocoumarin ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Chroman-2-One, 1,2-Benzodihydropyrone
D1223-25G
D1223-500G
TCIAD1223-25GEA
215
SEK
TCIAD1223-25G
TCIAD1223-500G
Dihydrocoumarin ≥98.0%
Dihydrocoumarin
Formel:
C₉H₈O₂ MW: 148,16 g/mol Kokpunkt: 149°C |
MDL Number:
MFCD00006881 CAS nummer: 119-84-6 EINECS: 204-354-9 |
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...