2-Chloro-5-nitropyrimidine ≥98.0%
Leverantör: TCI
|
Fara
|
C2507-1G
C2507-5G
TCIAC2507-1GEA
910
SEK
TCIAC2507-1G
TCIAC2507-5G
2-Chloro-5-nitropyrimidine ≥98.0%
2-Chloro-5-nitropyrimidine
Formel:
C₄H₂ClN₃O₂ Storage Temperature: Frys |
MDL Number:
MFCD04117995 CAS nummer: 10320-42-0 EINECS: 233-703-8 |
|
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 107.0 to 111.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...