2-Chloro-5-ethylpyrimidine ≥98.0%
Leverantör: TCI EUROPE
Varning
C1754-1G C1754-5G
TCIAC1754-1GEA 245 SEK
TCIAC1754-1G TCIAC1754-5G
2-Chloro-5-ethylpyrimidine ≥98.0%
2-Chloro-5-ethylpyrimidine
Formel: C₆H₇ClN₂
MW: 142,59 g/mol
Kokpunkt: 86°C
MDL Number: MFCD00799503
CAS nummer: 111196-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=749a0a8b3e0d02aea8599f0c24227fe5f7c4fcd2&vid=749a0a8b3e0d02aea8599f0c24227fe5f7c4fcd2&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid to slightly cloudy liquid
Purity (GC) min. 98.0 %