2,3-Dimethylhydroquinone ≥98.0%
Leverantör: TCI
|
Varning
|
D3877-25G
D3877-5G
TCIAD3877-25GEA
4025
SEK
TCIAD3877-25G
TCIAD3877-5G
2,3-Dimethylhydroquinone ≥98.0%
2,3-Dimethylhydroquinone
Formel:
C₈H₁₀O₂ Smältpunkt: 224°C |
MDL Number:
MFCD00009997 CAS nummer: 608-43-5 |
|
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...