1-Bromo-3-methoxypropane ≥98.0%

Leverantör: TCI
Varning

B3499-25G B3499-5G
TCIAB3499-25GEA 855 SEK
TCIAB3499-25G TCIAB3499-5G
1-Bromo-3-methoxypropane ≥98.0%
1-Bromo-3-methoxypropane
Formel: C₄H₉BrO
Kokpunkt: 132°C
MDL Number: MFCD02258473
CAS nummer: 36865-41-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=722eb39a200430f48514d2938397c5bb4f40f705&vid=722eb39a200430f48514d2938397c5bb4f40f705&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR