1-Butyl-1-methylpyrrolidinium chloride ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1-Butyl-1-methylpyrrolidin-1-ium chloride

B3358-25G B3358-5G
TCIAB3358-25GEA 1725 SEK
TCIAB3358-25G TCIAB3358-5G
1-Butyl-1-methylpyrrolidinium chloride ≥98.0%
1-Butyl-1-methylpyrrolidinium chloride
Formel: C₉H₂₀ClN
MW: 177,72 g/mol
MDL Number: MFCD08458922
CAS nummer: 479500-35-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f71365c80476a2ec1bcdff87ff2b5f17cb2b9305&vid=f71365c80476a2ec1bcdff87ff2b5f17cb2b9305&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR