p-Phenylenediacetonitrile ≥98.0%

Leverantör: TCI
Fara

Synonyms: 2,2'-(1,4-Phenylene)diacetonitrile, 1,4-Phenylenediacetonitrile

X0061-25G X0061-5G
TCIAX0061-25GEA 3025 SEK
TCIAX0061-25G TCIAX0061-5G
p-Phenylenediacetonitrile ≥98.0%
p-Phenylenediacetonitrile
Formel: C₁₀H₈N₂
MDL Number: MFCD00001923
CAS nummer: 622-75-3
EINECS: 210-751-8
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb9b594998a445ccfc328b131d3809d7245f56ba&vid=bb9b594998a445ccfc328b131d3809d7245f56ba&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 99.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR