Methyl carbazate ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Methyl hydrazinecarboxylate, Methyl hydrazinocarboxylate
C1098-25G
C1098-500G
TCIAC1098-25GEA
215
SEK
TCIAC1098-25G
TCIAC1098-500G
Methyl carbazate ≥98.0% (by GC, titration analysis)
Methyl carbazate
Formel:
C₂H₆N₂O₂ MW: 90,08 g/mol Kokpunkt: 108°C |
MDL Number:
MFCD00007594 CAS nummer: 6294-89-9 EINECS: 228-560-3 UN: 2811 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 72.0 to 74.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...