(±)-Ethyl-2-chloropropionate ≥97.0%

Leverantör: TCI EUROPE
Varning

C0790-100ML C0790-25ML
TCIAC0790-100MLEA 1850 SEK
TCIAC0790-100ML TCIAC0790-25ML
(±)-Ethyl-2-chloropropionate ≥97.0%
(±)-Ethyl-2-chloropropionate
Formel: C₅H₉ClO₂
MW: 136,58 g/mol
Kokpunkt: 147°C
Flampunkt: 38°C
MDL Number: MFCD00000864
CAS nummer: 535-13-7
EINECS: 208-610-0
UN: 2935
ADR: 3,III
Merck Index: 14,03785
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6650807867da6b3279512042204db3edd8e5d47a&vid=6650807867da6b3279512042204db3edd8e5d47a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR