(±)-2-Chloropropionyl chloride ≥95.0% (by GC, titration analysis)

Leverantör: TCI
Fara

C1114-25G C1114-500G C1114-100G
TCIAC1114-25GEA 255 SEK
TCIAC1114-25G TCIAC1114-500G TCIAC1114-100G
(±)-2-Chloropropionyl chloride ≥95.0% (by GC, titration analysis)
(±)-2-Chloropropionyl chloride
Formel: C₃H₄Cl₂O
Kokpunkt: 111°C
Flampunkt: 31°C
MDL Number: MFCD00000862
CAS nummer: 7623-09-8
EINECS: 231-540-7
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a3d5e068e70970bc42f22033957a1b416e342f9&vid=9a3d5e068e70970bc42f22033957a1b416e342f9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR