5-Methoxy-2-methylbenzothiazole ≥98.0%

Leverantör: TCI EUROPE

M1281-25G M1281-5G
TCIAM1281-25GEA 4275 SEK
TCIAM1281-25G TCIAM1281-5G
5-Methoxy-2-methylbenzothiazole ≥98.0%
5-Methoxy-2-methylbenzothiazole
Formel: C₉H₉NOS
MW: 179,24 g/mol
Kokpunkt: 135°C
MDL Number: MFCD00082664
CAS nummer: 2941-69-7
EINECS: 220-929-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d567a36c03e2a211e852cf38b5cb1e9dbea0a40&vid=9d567a36c03e2a211e852cf38b5cb1e9dbea0a40&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR