(1R)-(-)-Camphorquinone ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: (1R,4S)-1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione

C1482-1G C1482-5G
TCIAC1482-1GEA 495 SEK
TCIAC1482-1G TCIAC1482-5G
(1R)-(-)-Camphorquinone ≥98.0%
(1R)-(-)-Camphorquinone
Formel: C₁₀H₁₄O₂
MW: 166,22 g/mol
MDL Number: MFCD00082863
CAS nummer: 10334-26-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e89d10c89e5d6ce19f47fe59b7c5c0693c565692&vid=e89d10c89e5d6ce19f47fe59b7c5c0693c565692&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 200.0 to 204.0 deg-C
Specific rotation [a]20/D -95.0 to -105.0 deg (C=1,toluene)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR