(±)-Ethyl-2,3-dibromopropionate ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Ethyl 2,3-Dibromopropionate

D1992-25G D1992-500G
TCIAD1992-25GEA 215 SEK
TCIAD1992-25G TCIAD1992-500G
(±)-Ethyl-2,3-dibromopropionate ≥98.0%
(±)-Ethyl-2,3-dibromopropionate
Formel: C₅H₈Br₂O₂
MW: 259,93 g/mol
Kokpunkt: 82°C
Flampunkt: 91°C
MDL Number: MFCD00000212
CAS nummer: 3674-13-3
EINECS: 222-941-8
UN: 2922
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=364b63c7dd5ca7f591c0c70f6a341d7d8ff8e788&vid=364b63c7dd5ca7f591c0c70f6a341d7d8ff8e788&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR